Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid NTD Protein (His Tag)
Supplier:  Sino Biological 40588V07E10100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid NTD (YP_009724397.2) (Thr49-Gly175) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6892