Learn More
Sino Biological Recombinant Anti-IL-13 R alpha 1/IL13RA1 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10943R038P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-13RA1 / CD213a1 (rh IL-13RA1 / CD213a1; Catalog#10943-H08H; NP_001551.1; Met 1-Thr 343) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-5038