Learn More
Sino Biological Human IL13Ra1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10943MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL13RA1 extracellular domain (rh IL13RA1; Catalog#10943-H08H; NP_001551.1; Met 1-Thr 343). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2209