Learn More
Sino Biological Anti-IL-13 R alpha 1/IL13RA1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10943MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL13RA1 / IL-13RA1 (rh IL13RA1 / IL-13RA1; Catalog#10943-H08H; NP_001551.1; Met1-Thr343). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5042