Learn More
Sino Biological Anti-IL-13 R alpha 1/IL13RA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10943T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL13RA1/IL-13RA1 (rh IL13RA1/IL-13RA1; Catalog#10943-H08H; NP_001551.1; Met1-Thr343). IL13RA1/IL-13RA1 specific IgG was purified by Human IL13RA1/IL-13RA1 affinity chromatography.
Catalog No. 50-255-5034