Learn More
Sino Biological Recombinant Human IL-13 R alpha 2/IL13RA2 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10350H41CB100
Encompass_Preferred
A DNA sequence encoding the Human IL13Ra2 (NP_000631.1)(Met1-Arg343) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5361