Learn More
Sino Biological Recombinant Human IL-13 R alpha 2/IL13RA2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10350H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL13Rα2 (NP_000631.1) (Met1-Leu342) was expressed with the a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5362