Learn More
Proteintech Group Inc Human IL15 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0010296T
Encompass_Preferred
Interleukin 15 (IL15) is a cytokine important for the development, maturation, and function of many cells of the immune system including NK, NKT, gammadeltaT, and CD8(+) T cells. Its expression affects homeostasis and function of B cells through NK cell-derived IFNG. IL-15 binds a trimeric IL-15 receptor (IL-15R) complex formed by a specific receptor chain (IL-15Rα) combined with theβandγchains of the IL-2R, acting as signal transducing components. High levels of IL15 expression have been associated with the pathogenesis of autoimmune and inflammatory diseases like Crohn’s disease, psoriasis, leukemias, rheumatoid arthritis (RA) and graft rejection, resulting in the IL15 system being a target of interest for the treatment of these diseases
Catalog No. 50-197-4062