Learn More
Sino Biological Human IL15 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10428H49HB20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-15 / IL15 / Interleukin 15 (rh IL-15 / IL15 / Interleukin 15; Catalog#10360-H07E; NP_000576.1; Asn49-Ser162).
Catalog No. 50-237-4224