Learn More
Sino Biological Recombinant Anti-IL-15 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10360R111H20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-15 / IL15 / Interleukin 15 (rh IL-15 / IL15 / Interleukin 15; Catalog#10360-H07E; NP_000576.1; Asn49-Ser162) and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-5351