Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL15 (P40933) Recombinant Protein

Catalog No. 89948656 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-656 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-656 Supplier Abnova™ Supplier No. P4407
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene is a cytokine that regulates T and natural killer cell activation and proliferation. This cytokine and interleukine 2 share many biological activities. They are found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between this cytokine and IL2. This cytokine induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. Studies of the mouse counterpart suggested that this cytokine may increase the expression of apoptosis inhibitor BCL2L1/BCL-x(L), possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported. [provided by RefSeq]

Sequence: MNWVNVISDLKKIEDLIQSMHIDATLYTESDVHPSCKVTAMKCFLLELQVISLESGDASIHDTVENLIILANNSLSSNGNVTESGCKECEELEEKNIKEFLQSFVHIVQMFINTS

Specifications

Accession Number P40933
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 12.8kDa
Name IL15 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MNWVNVISDLKKIEDLIQSMHIDATLYTESDVHPSCKVTAMKCFLLELQVISLESGDASIHDTVENLIILANNSLSSNGNVTESGCKECEELEEKNIKEFLQSFVHIVQMFINTS
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias IL-15/MGC9721
Common Name IL15
Gene Symbol IL15
Biological Activity The activity is determined by the dose-dependant stimulation of the proliferation of mouse CTLL-2 cells and is typcially less than 0.5ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.