Learn More
Sino Biological Human IL15RA Protein 20ug (Sino Biological)
Supplier:  Sino Biological 29736H41HB20
Encompass_Preferred
A DNA sequence encoding the human IL15RA (NP_002180.1) (Met1-Thr205) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-3689