Learn More
Sino Biological Recombinant Human IL-15 R alpha/IL15RA Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 18366H02H150
Encompass_Preferred
A DNA sequence encoding the human IL15RA (NP_002180.1) (Ile31-Thr205) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4002