Learn More
Sino Biological Human IL16 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10944R373100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-16 (rh IL-16; Catalog#10944-H07E; AAC12732.1; Pro 2-Ser 130).
Catalog No. 50-237-2219