Learn More
Sino Biological Human IL17A ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502373484
Encompass_Preferred
Each vial contains 41 ng of recombinant Human IL17A/IL-17A/IL17. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 75 pg/mL is recommended.
Catalog No. 50-237-3484 Supplier Sino Biological Supplier No. SEK120475
Each vial contains 41 ng of recombinant Human IL17A/IL-17A/IL17. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 75 pg/mL is recommended.