Learn More
Sino Biological Recombinant Human IL17A & IL17F Heterodimer Protein (His Tag) 1
Supplier:  Sino Biological CT047H08H1
Encompass_Preferred
A DNA sequence encoding the human IL17A (NP_002181.1) (Met1-Ala155) and the mature form of human IL17F (NP_443104.1) (Arg31-Gln163) linked by a polypeptide linker was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6843