Learn More
Sino Biological Human IL17A & IL17F Protein 20ug (Sino Biological)
Supplier:  Sino Biological CT047HNAE20
Encompass_Preferred
A DNA sequence encoding the human IL17A (Q16552) (Ile20-Ala155) was expressed, constructed the plasmid 1; the human IL17F (NP_443104.1) (Arg31-Gln163) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-237-2806