Learn More
Sino Biological Recombinant Human IL-17RB Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 13091H08H1
Encompass_Preferred
A DNA sequence encoding the human IL17BR (NP_061195.2) (Met1-Gly289) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6595