Learn More
Sino Biological Recombinant Human IL-17F Protein (His & AVI Tag), Biotinylated, HPLC-verified
Catalog No. 502993375
Encompass_Preferred
A DNA sequence encoding the Human IL17F (NP_443104.1) (Arg31-Gln163) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3375 Supplier Sino Biological Supplier No. 11855H56H1B20
A DNA sequence encoding the Human IL17F (NP_443104.1) (Arg31-Gln163) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.