Learn More
Sino Biological Anti-IL-17F Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11855MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL17F (rh IL17F; Catalog#11855-H07H; NP_443104.1; Arg31-Gln163). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4515