Learn More
Sino Biological Anti-IL-17F Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310466T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human IL17F fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3788