Learn More
Sino Biological Human IL17F PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11855T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL17F (rh IL17F; Catalog#11855-HNAE; AAH70124.1; Arg31-Gln163). IL17F specific IgG was purified by Human IL17F affinity chromatography.
Catalog No. 50-237-1751