Learn More
Sino Biological Recombinant Human IL-17F Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11855H07HB20
Encompass_Preferred
A DNA sequence encoding the human IL17F (NP_443104.1) (Arg31-Gln163) was expressed, with a polyhistidine tag at the N-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4517