Learn More
Sino Biological Recombinant Human CD200 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10886H08H1
Encompass_Preferred
A DNA sequence encoding the human CD200 (NP_005935.4) extracellular domain (Met 1-Gly 232) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-3577