Learn More
Sino Biological Anti-IL-17RA Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10895MM06A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL17RA/IL-17RA/CD217 (rh IL17RA/IL-17RA/CD217; Catalog#10895-H08H; NP_055154.3; Met1-Trp320) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1173