Learn More
Sino Biological Recombinant Anti-IL-17RA Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10895R004F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL17RA/IL-17RA/CD217 (rh IL17RA/IL-17RA/CD217; Catalog#10895-H08H; NP_055154.3; Met1-Trp320) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8701