Learn More
Sino Biological Anti-IL-17RB Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13091MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL17BR / IL17RB / IL-17 Receptor B (rh IL17BR / IL17RB / IL-17 Receptor B; Catalog#13091-H08H; NP_061195.2; Met1-Gly289). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4319