Learn More
Sino Biological Anti-IL-17RB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13091T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL17BR / IL17RB / IL-17 Receptor B (rh IL17BR / IL17RB / IL-17 Receptor B; Catalog#13091-H08H; NP_061195.2; Met1-Gly289). IL17BR / IL17RB / IL-17 Receptor B specific IgG was purified by Human IL17BR / IL17RB / IL-17 Receptor B affinity chromatography.
Catalog No. 50-255-4318