Learn More
Sino Biological Anti-IL-17RC Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11747MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL17RC (rh IL17RC; Catalog#11747-H08B; NP_116121.2; Met 1-Ala 454). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4584