Learn More
Sino Biological Anti-IL-17RC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11747RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL17RC (rh IL17RC; Catalog#11747-H08B; NP_116121.2; Met 1-Ala 454). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4582