Learn More
Sino Biological Recombinant Human IL-17RC Protein (hFc Tag) 1
Supplier:  Sino Biological 11747H02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL17RC isoform 3 (NP_116121.2) (Met 1-Ala 454) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7264