Learn More
Sino Biological Anti-IL17RD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10507RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL17Rd (rhIL17Rd; Catalog#10507-H08H; NP_060033.3; Met 1-Arg 299). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8169