Learn More
Sino Biological Human IL18 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502370877
Encompass_Preferred
Each vial contains 114 ng of recombinant Human IL18/IL-18/Interleukin 18. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2500 pg/mL is recommended.
Catalog No. 50-237-0877 Supplier Sino Biological Supplier No. SEK101195
Each vial contains 114 ng of recombinant Human IL18/IL-18/Interleukin 18. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2500 pg/mL is recommended.