Learn More
Sino Biological Human IL18BP ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370519
Encompass_Preferred
Each vial contains 47 ng of recombinant Human IL18BP / IL18BPa. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-0519 Supplier Sino Biological Supplier No. SEKA1035715
Each vial contains 47 ng of recombinant Human IL18BP / IL18BPa. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.