Learn More
Sino Biological Anti-IL-18Rα Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 11102MM05H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL18R1 (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met1-Arg329). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8256