Learn More
Sino Biological Anti-IL-18Rα Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11102MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL18R1 / CD218a (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met 1-Arg 329). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2238