Learn More
Sino Biological Anti-IL-18Rα Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11102MM17F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL18R1 (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met1-Arg329) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8268