Learn More
Sino Biological Anti-IL-18Rα Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11102RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL18R1 / CD218a (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met 1-Arg 329). IL18R1 specific IgG was purified by human IL18R1 affinity chromatography.
Catalog No. 50-237-4495