Learn More
Sino Biological Human IL18R1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11102RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL18R1 / CD218a (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met 1-Arg 329). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-0569