Learn More
Sino Biological Recombinant Human IL-18Rα Protein (ECD, His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 11102H08HB20
Encompass_Preferred
A DNA sequence encoding the Human IL18R1 (NP_003846.1) (Met1-Arg329) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-240-7785