Learn More
Sino Biological Recombinant Human IL-18R beta / IL-18RAP Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10176H49HB20
Encompass_Preferred
A DNA sequence encoding the Human IL18RAP (NP_003844.1)(Met1-Gly357) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8135