Learn More
Sino Biological Anti-IL-19 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13089MM1120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL19 / Interleukin-19 (rh IL19 / Interleukin-19; Catalog#13089-H08H; AAF06663.1; Met1-Ala177). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8874