Learn More
BIOHIPPO Human IL19 Microsample
SDP
Catalog No. 5000314252
Encompass
Human IL19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL19 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14252 Supplier BIOHIPPO Supplier No. BHE15211794
Human IL19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL19 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color