Learn More
Sino Biological Anti-IL-19 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13089RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL19 / Interleukin-19 (rh IL19; Catalog#13089-H08H; AAF06663.1; Met1-Ala177). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9385