Learn More
Sino Biological Human IL1A Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10128HNCE20
Encompass_Preferred
A DNA sequence encoding the human IL1A (NP_000566.3) (Ser113-Ala271) was expressed.
Catalog No. 50-237-3244