Learn More
Sino Biological Anti-IL-1 beta Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10139MM48100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1 beta (rh IL-1 beta; Catalog#10139-HNAE; NP_000567.1; Ala117-Ser269). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1176