Learn More
Sino Biological Human IL1B MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10139M20120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1 beta / IL1B (rh IL-1 beta / IL1B; Catalog#10139-H07E; NP_000567.1; Met1-Ser269). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0800