Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310202T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human IL-1 beta fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8463