Learn More
Sino Biological Recombinant Human IL-1 beta Protein (pro form, His Tag)
Supplier:  Sino Biological 10139H07E1
Encompass_Preferred
A DNA sequence encoding the pro form of human IL1β (NP_000567.1, with mutation Glu 6 Lys) (Met 1-Ser 269) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-5454