Learn More
Sino Biological Human IL1B Protein 20ug (Sino Biological)
Supplier:  Sino Biological GMP10139HNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1B (NP_000567.1) (Ala 117-Ser 269) was expressed with a N-terminal Met.
Catalog No. 50-237-2991